Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560185
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560185 Supplier Invitrogen™ Supplier No. PA560185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKRCRSAQVL HLYGATYSAD GEDRARCSAG AHTLLVQLPE RTVLLDAYSE HLAAALCTNP NLIELSLYRN ALGS Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 53%.

NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP12, also known as Monarch-1, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. NALP12 is thought to act as an attenuating factor of inflammation by suppressing inflammatory responses such as NF-kappa-B activation by TLR-signaling molecules MyD88, IRAK-1, TRAF6 and RIPK1 in activated monocytes. Recent evidence suggests that mutations in NALP12 result in hereditary periodic fever syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLRP12
Gene Accession No. P59046
Gene Alias CLR19.3; FCAS2; monarch 1; monarch-1; NACHT, leucine rich repeat and PYD containing 12; NACHT, LRR and PYD containing protein 12; NACHT, LRR and PYD domains-containing protein 12; NALP12; NLR family pyrin domain containing 12; NLR family, pyrin domain containing 12; Nlrp12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PAN6; PYPAF7; PYRIN-containing APAF1-like protein 7; regulated by nitric oxide; RNO; RNO2
Gene Symbols NLRP12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NALP12. Recombinant protein control fragment (Product #RP-99104).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.