Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NALP12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572911
Description
NALP12 Polyclonal Antibody for Western Blot, IHC (F), IHC (P), IP
NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP12, also known as Monarch-1, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. NALP12 is thought to act as an attenuating factor of inflammation by suppressing inflammatory responses such as NF-kappa-B activation by TLR-signaling molecules MyD88, IRAK-1, TRAF6 and RIPK1 in activated monocytes. Recent evidence suggests that mutations in NALP12 result in hereditary periodic fever syndromes.
Specifications
NALP12 | |
Polyclonal | |
Unconjugated | |
NLRP12 | |
CLR19.3; FCAS2; monarch 1; monarch-1; NACHT, leucine rich repeat and PYD containing 12; NACHT, LRR and PYD containing protein 12; NACHT, LRR and PYD domains-containing protein 12; NALP12; NLR family pyrin domain containing 12; NLR family, pyrin domain containing 12; Nlrp12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PAN6; PYPAF7; PYRIN-containing APAF1-like protein 7; regulated by nitric oxide; RNO; RNO2 | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P59046 | |
NLRP12 | |
A synthetic peptide corresponding to amino acids 190-208 of human NALP12/PYPAF7/Monarch-1 was used as immunogen (NP_653288). | |
RUO | |
91662 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction