Learn More
Invitrogen™ NALP12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589879
Description
Positive Samples: Rat spleen; Cellular Location: Cytoplasm Immunogen sequence: WLKICRLTAA ACDELASTLS VNQSLRELDL SLNELGDLGV LLLCEGLRHP TCKLQTLRLG ICRLGSAACE GLSVVLQANH NLRELDLSFN DLGDWGLWLL AEGLQHPACR LQKLWLDSCG LTAKACENLY FTLGINQTLT DLYLTNNALG DTGVRLLCKR LSHPGCKLRV LWLFGMDLNK MTHSRLAALR VTKPYLDIGC.
NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP12, also known as Monarch-1, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. NALP12 is thought to act as an attenuating factor of inflammation by suppressing inflammatory responses such as NF-kappa-B activation by TLR-signaling molecules MyD88, IRAK-1, TRAF6 and RIPK1 in activated monocytes. Recent evidence suggests that mutations in NALP12 result in hereditary periodic fever syndromes.
Specifications
NALP12 | |
Polyclonal | |
Unconjugated | |
NLRP12 | |
CLR19.3; FCAS2; monarch 1; monarch-1; NACHT, leucine rich repeat and PYD containing 12; NACHT, LRR and PYD containing protein 12; NACHT, LRR and PYD domains-containing protein 12; NALP12; NLR family pyrin domain containing 12; NLR family, pyrin domain containing 12; Nlrp12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PAN6; PYPAF7; PYRIN-containing APAF1-like protein 7; regulated by nitric oxide; RNO; RNO2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
292541, 91662 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.07 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P59046 | |
NLRP12 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 862-1061 of human NLRP12 (NP_6532881). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.