Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521027
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521027 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521027 Supplier Invitrogen™ Supplier No. PA521027
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-21027 can be used with blocking peptide PEP-1141.

NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP12, also known as Monarch-1, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. NALP12 is thought to act as an attenuating factor of inflammation by suppressing inflammatory responses such as NF-kappa-B activation by TLR-signaling molecules MyD88, IRAK-1, TRAF6 and RIPK1 in activated monocytes. Recent evidence suggests that mutations in NALP12 result in hereditary periodic fever syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP12
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NLRP12
Gene Accession No. E9Q5R7, P59046
Gene Alias CLR19.3; FCAS2; monarch 1; monarch-1; NACHT, leucine rich repeat and PYD containing 12; NACHT, LRR and PYD containing protein 12; NACHT, LRR and PYD domains-containing protein 12; NALP12; NLR family pyrin domain containing 12; NLR family, pyrin domain containing 12; Nlrp12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PAN6; PYPAF7; PYRIN-containing APAF1-like protein 7; regulated by nitric oxide; RNO; RNO2
Gene Symbols NLRP12
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human NALP12.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292541, 378425, 91662
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.