Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563859 Supplier Invitrogen™ Supplier No. PA563859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLPYLMALDQ YQLEEFKLCL EPQQLMDFWS APQGHFPRIP WANLRAADPL NLSFLLDEHF PKGQAWKVVL GIFQTMNLTS LCEKVRAEMK ENVQTQELQD PTQED Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 28%.

NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP13 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. While little is known about the function of NALP13, it has been suggested that it may be implicated in the activation of proinflammatory caspases via inflammasomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLRP13
Gene Accession No. Q86W25
Gene Alias CLR19.7; NACHT, leucine rich repeat and PYD containing 13; NACHT, LRR and PYD containing protein 13; NACHT, LRR and PYD domains-containing protein 13; NALP13; NLR family pyrin domain containing 13; NLR family, pyrin domain containing 13; NLRP13; NOD14; Nucleotide-binding oligomerization domain protein 14; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 13; PAN13
Gene Symbols NLRP13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NALP13. Recombinant protein control fragment (Product #RP-99157).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.