Learn More
Invitrogen™ NALP13 Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-21028 can be used with blocking peptide PEP-1142.
Specifications
Specifications
| Antigen | NALP13 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NLRP13 |
| Gene Accession No. | Q86W25 |
| Gene Alias | CLR19.7; NACHT, leucine rich repeat and PYD containing 13; NACHT, LRR and PYD containing protein 13; NACHT, LRR and PYD domains-containing protein 13; NALP13; NLR family pyrin domain containing 13; NLR family, pyrin domain containing 13; NLRP13; NOD14; Nucleotide-binding oligomerization domain protein 14; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 13; PAN13 |
| Gene Symbols | NLRP13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.