Learn More
Invitrogen™ NALP2 Monoclonal Antibody (1B10)

Description
Peptide Sequence: MVSSAQMGFN LQALLEQLSQ DELSKFKYLI TTFSLAHELQ KIPHKEVDKA DGKQLVEVLT THCDSYWVEM ASLQVFEKMH RMDLSERAKD EVREAALKSF NKRKPLSLGI TRKERPPLDV DEMLERFKTE AQAFTETKGN VICLGKEVFK GKKPDKDNRC RYILKTKFRE MWKSWPGDSK EVQVMAERYK MLIPFSNPRV LPGPFSYTVV LYGPAGLGKT TLAQKLMLDW AEDNLIHKFK YAFYLSCREL SRLGPCSFAE LVFRDWPELQ DDIPHILAQA RKILFVIDGF DELGAAPGAL IEDICGDWEK KKPVPVLLGS LLNRVMLPKA ALLVTTRPRA LRDLRILAEE PIYIRVEGFL EEDRRAYFLR HFGDEDQAMR AFELMRSNAA LFQLGSAPAV CWIVCTTLKL QMEKGEDPVP TCLTRTGLFL RFLCSRFPQG AQLRGALRTL SLLAAQGLWA QTSVLHREDL ERLGVQESDL RLFLDGDILR QDRVSKGCYS FIHLSFQQFL TALFYTLEKE EEEDRDGHTW DIGDVQKLLS GVERLRNPDL IQAGYYSFGL ANEKRAKELE ATFGCRMSPD IKQELLRCDI SCKGGHSTVT DLQELLGCLY ESQEEELVKE VMAQFKEISL HLNAVDVVPS SFCVKHCRNL QKMSLQVIKE NLPENVTASE SDAEVERSQD DQHMLPFWTD LCSIFGSNKD LMGLAINDSF LSASLVRILC EQIASDTCHL QRVVFKNISP ADAHRNLCLA LRGHKTVTYL TLQGNDQDDM FPALCEVLRH PECNLRYLGL VSCSATTQQW ADLSLALEVN QSLTCVNLSD NELLDEGAKL LYTTLRHPKC FLQRLS.
Specifications
Specifications
| Antigen | NALP2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1B10 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NLRP2 |
| Gene Accession No. | Q9NX02 |
| Gene Alias | CLR19.9; E330007A02Rik; FLJ20510; LOC684545; NACHT, leucine rich repeat and PYD containing 2; NACHT, LRR and PYD containing protein 2; NACHT, LRR and PYD domains-containing protein 2; NALP2; NBS1; NLR family pyrin domain containing 2; NLR family, pyrin domain containing 2; NLRP2; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 2; nucleotide-binding site protein 1; PAN1; PYPAF2; PYRIN domain and NACHT domain-containing protein 1; PYRIN-containing APAF1-like protein 2; similar to NACHT, leucine rich repeat and PYD containing 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.