Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598859 Supplier Invitrogen™ Supplier No. PA598859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NALP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

NALP2 belongs to a family of cytoplasmic proteins that have been implicated in cell responses to apoptotic and inflammatory stimuli. Unlike the prototypical NALP protein NALP1, NALP2 only contains a NACHT domain, leucine rich repeat (LRR), and pyrin-containing domain (PYD). This protein interacts with the adapter protein ASC in addition to CARD8 and caspase-1 to form an inflammasome with high proIL-1-b-processing activity and is thought to function as a modulator of NF-kappa-B and procaspase-1 activation in macrophages. It has also been suggested that NALP2, in addition to other NALP family members, can act as innate sensors of pathogens in a manner similar to the toll-like receptors (TLRs). At least two alternatively spliced transcript variants encoding distinct isoforms have been found for NALP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NLRP2
Gene Accession No. Q9NX02
Gene Alias CLR19.9; E330007A02Rik; FLJ20510; LOC684545; NACHT, leucine rich repeat and PYD containing 2; NACHT, LRR and PYD containing protein 2; NACHT, LRR and PYD domains-containing protein 2; NALP2; NBS1; NLR family pyrin domain containing 2; NLR family, pyrin domain containing 2; NLRP2; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 2; nucleotide-binding site protein 1; PAN1; PYPAF2; PYRIN domain and NACHT domain-containing protein 1; PYRIN-containing APAF1-like protein 2; similar to NACHT, leucine rich repeat and PYD containing 2
Gene Symbols NLRP2
Host Species Rabbit
Immunogen Recombinant Human NACHT, LRR and PYD domains-containing protein 2 protein (1-220AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55655
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.