Learn More
Specifications
Specifications
| Gene ID (Entrez) | 147945 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56156 |
| Gene Alias | Cancer/testis antigen 58, CLR19.5, CT58PYRIN and NACHT-containing protein 2, FLJ32126, NACHT, leucine rich repeat and PYD containing 4, NACHT, LRR and PYD domains-containing protein 4, NALP4NACHT, LRR and PYD containing protein 4, NLR family, pyrin domain containing 4, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 4, PAN2Ribonuclease inhibitor 2, PYPAF4PAAD and NACHT-containing protein 2, RNH2PYRIN-containing APAF1-like protein 4 |
| Gene Symbol | NLRP4 |
| Quantity | 0.1 mg |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
