Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NALP4 Polyclonal Antibody
NALP4 Polyclonal Antibody
Supplier: Thermo Scientific PA572910
Description
NALP4 Polyclonal Antibody for Western Blot, IHC (P), IP

Specifications
NALP4 | |
Polyclonal | |
Unconjugated | |
NLRP4 | |
4930406H16Rik; cancer/testis antigen 58; CLR19.5; CT58; E330021B02; E330028A19Rik; NACHT, leucine rich repeat and PYD containing 4; NACHT, leucine rich repeat and PYD containing 4A; NACHT, leucine rich repeat and PYD containing 4E; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NACHT, LRR and PYD domains-containing protein 4A; NACHT, LRR and PYD domains-containing protein 4E; NACHT/LRR/pyrin domain-containing protein 4; NALP, epsilon; NALP4; Nalp4a; Nalp4e; NALP-epsilon; Nalp-eta; Nalp-ita; NLR family pyrin domain containing 4; NLR family, pyrin domain containing 4; NLR family, pyrin domain containing 4A; NLR family, pyrin domain containing 4E; Nlrp4; Nlrp4a; Nlrp4e; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; RGD1561918; RGD1563529; Ribonuclease inhibitor 2; RNH2 | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
Whole serum with 0.02% sodium azide | |
Q66X19, Q8BU40, Q96MN2 | |
NLRP4, Nlrp4a, Nlrp4e | |
A synthetic peptide corresponding to amino acids 139-157 of human NALP4/PAN2/PYPAF4 was used as immunogen; GenBank no. gi 17064172 gb AAL35293.1 AF442488_1. | |
RUO | |
147945, 243880, 446099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction