Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139855 Supplier Invitrogen™ Supplier No. PA5139855
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FTLTKLSRDD IRSLCDALNY PAGNVKELAL VNCHLSPIDC EVLAGLLTNN KKLTYLNVSC NQLDTGVPLL CEALCSPDTV LVYLMLAFCH LS Highest antigen sequence indentity to the following orthologs: Rat - 48%, Mouse - 47%.

NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP4, also known as PYRIN-containing APAF1-like protein 4, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. In transfected 293 cells, NALP4 suppressed NF-kappa-B induction by TNF-a and IL-1-b, suggesting NALP4 operates at a point of convergence in these two signaling pathways. NALP4 also suppressed NF-kappa-B induction resulting from overexpression of several adapter proteins and protein kinases involved in these pathways, including TRAF2, TRAF6, RIP, and IRAK2, as well as the IKK-alpha and IKK-beta, suggesting that NALP4 is critical in modulating NF-kappa-B activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLRP4
Gene Accession No. Q96MN2
Gene Alias 4930406H16Rik; cancer/testis antigen 58; CLR19.5; CT58; E330021B02; E330028A19Rik; NACHT, leucine rich repeat and PYD containing 4; NACHT, leucine rich repeat and PYD containing 4A; NACHT, leucine rich repeat and PYD containing 4E; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NACHT, LRR and PYD domains-containing protein 4A; NACHT, LRR and PYD domains-containing protein 4E; NACHT/LRR/pyrin domain-containing protein 4; NALP, epsilon; NALP4; Nalp4a; Nalp4e; NALP-epsilon; Nalp-eta; Nalp-ita; NLR family pyrin domain containing 4; NLR family, pyrin domain containing 4; NLR family, pyrin domain containing 4A; NLR family, pyrin domain containing 4E; Nlrp4; Nlrp4a; Nlrp4e; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; RGD1561918; RGD1563529; Ribonuclease inhibitor 2; RNH2
Gene Symbols NLRP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NLRP4. Recombinant protein control fragment (Product #RP-109015).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147945
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.