Learn More
NALP4 Rabbit anti-Human, Polyclonal, Bioss
SDP
Rabbit Polyclonal Antibody
Supplier:  Bioss BS6855R
Encompass_Preferred
Catalog No. 50-198-7550
Only null left
Add to Cart
Description
Description
NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP4, also known as PYRIN-containing APAF1-like protein 4, has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. In transfected 293 cells, NALP4 suppressed NF-kappa-B induction by TNF-a and IL-1-b, suggesting NALP4 operates at a point of convergence in these two signaling pathways. NALP4 also suppressed NF-kappa-B induction resulting from overexpression of several adapter proteins and protein kinases involved in these pathways, including TRAF2, TRAF6, RIP, and IRAK2, as well as the IKK-alpha and IKK-beta, suggesting that NALP4 is critical in modulating NF-kappa-B activity.
Specifications
Specifications
NALP4
Polyclonal
Unconjugated
NLRP4
4930406H16Rik; Cancer/testis antigen 58; CLR19.5; CT58; E330021B02; E330028A19Rik; NACHT, leucine rich repeat and PYD containing 4; NACHT, leucine rich repeat and PYD containing 4A; NACHT, leucine rich repeat and PYD containing 4E; NACHT, LRR and PYD containing protein 4; NACHT, LRR and PYD domains-containing protein 4; NACHT, LRR and PYD domains-containing protein 4A; NACHT, LRR and PYD domains-containing protein 4E; NACHT/LRR/pyrin domain-containing protein 4; NALP, epsilon; NALP4; Nalp4a; Nalp4e; NALP-epsilon; Nalp-eta; NALP-ita; NLR family pyrin domain containing 4; NLR family, pyrin domain containing 4; NLR family, pyrin domain containing 4A; NLR family, pyrin domain containing 4E; Nlrp4; Nlrp4a; Nlrp4e; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 4; PAAD and NACHT-containing protein 2; PAN2; PYPAF4; PYRIN and NACHT-containing protein 2; PYRIN-containing APAF1-like protein 4; Ribonuclease inhibitor 2; RNH2
Rabbit
Protein A
RUO
147945
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q96MN2
NLRP4
KLH conjugated synthetic peptide derived from human NALP4.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.