Learn More
Invitrogen™ NALP5 Polyclonal Antibody

Description
NALP5 Polyclonal Antibody for ICC/IF
Specifications
Specifications
| Antigen | NALP5 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | NLRP5 |
| Gene Accession No. | P59047, Q9R1M5 |
| Gene Alias | CLR19.8; MATER; Mater protein; Mater protein homolog; maternal antigen that embryos require; NACHT, leucine rich repeat and PYD containing 5; NACHT, LRR and PYD domains-containing protein 5; Nalp5; NLR family pyrin domain containing 5; NLR family, pyrin domain containing 5; NLRP5; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 5; Ooplasm-specific protein 1; OP1; PAN11; PYPAF8 |
| Gene Symbols | NLRP5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.