Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NALP6 Antibody (Clint-1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311675V
Description
NALP6 Monoclonal specifically detects NALP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
NALP6 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
>95%, by SDS-PAGE | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Clint-1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
Recombinant human NALP6 (NACHT domain). | |
0.1 mL | |
Immunology, Inflammation | |
171389 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction