Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NALP8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563381
Description
Immunogen sequence: EDLNVGETQV NLEEGESGKI RRYKSNVMEK FFPIWDITTW PGNQRDFFYQ GVHRHEEYLP CLLLPKRPQG RQPKTVA Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 26%.
NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP8 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. It maps to chromosome 19q13.42 and is thought to be expressed in testis and ovary, suggesting that NALP8 may be involved in reproduction. Surprisingly, NALP8 seems to be universal in mammals except for rodents, suggesting that the gene may have been lost during evolution.
Specifications
NALP8 | |
Polyclonal | |
Unconjugated | |
NLRP8 | |
CLR19.2; NACHT, leucine rich repeat and PYD containing 8; NACHT, LRR and PYD containing protein 8; NACHT, LRR and PYD domains-containing protein 8; NALP8; NLR family pyrin domain containing 8; NLR family, pyrin domain containing 8; NLRP8; NOD16; Nucleotide-binding oligomerization domain protein 16; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 8; PAN4; PYRIN and NACHT-containing protein 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
126205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86W28 | |
NLRP8 | |
Recombinant protein corresponding to Human NALP8. Recombinant protein control fragment (Product #RP-101527). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction