Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NALP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560053
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560053 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560053 Supplier Invitrogen™ Supplier No. PA560053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITQLVRASVV QPQTLLTQVG IFMFGISTEE IVSMLETSFG FPLSKDLKQE ITQCLESLSQ CEADREAIAF QELFIGL Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 39%.

NOD6, also known as NALP9, is a member of the NALP family, a group of proteins that typically contain a NACHT domain, a NACHT-associated domain (NAD), a C-terminal leucine-rich repeat (LRR) region, and an N-terminal pyrin domain (PYD) and are involved in inflammation and innate immune defense. The bovine NOD6, which has 76% homology to its human counterpart, has been suggested to be an oocyte marker gene. In adult tissues, NALP9 mRNA is expressed exclusively in ovary and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NALP9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLRP9
Gene Accession No. Q7RTR0
Gene Alias CLR19.1; D7Ertd565e; NACHT, leucine rich repeat and PYD containing 9; NACHT, LRR and PYD containing protein 9; NACHT, LRR and PYD containing protein 9a; NACHT, LRR and PYD containing protein 9b; NACHT, LRR and PYD domains-containing protein 9; NACHT, LRR and PYD domains-containing protein 9A; NALP9; Nalp9a; Nalp9b; NALP-theta; NLR family pyrin domain containing 9; NLR family, pyrin domain containing 9; NLR family, pyrin domain containing 9A; NLR family, pyrin domain containing 9B; NLRP9; Nlrp9a; Nlrp9b; NOD6; Nucleotide-binding oligomerization domain protein 6; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 9; PAN12; PYRIN and NACHT-containing protein 12
Gene Symbols NLRP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NALP9. Recombinant protein control fragment (Product #RP-99186).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338321
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.