Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Benzoyl-L-argininamide Hydrochloride Monohydrate 98.0+%, TCI America™
SDP

Catalog No. B010225G
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 4299-03-0
Molecular Formula C13H20ClN5O2
Molecular Weight (g/mol) 313.786
MDL Number MFCD00035003
InChI Key PYZACNCNNFUUDO-PPHPATTJSA-N
Synonym s-n-1-amino-5-guanidino-1-oxopentan-2-yl benzamide hydrochloride, n-alpha-benzoyl-l-argininamide hydrochloride, 2s-5-carbamimidamido-2-phenylformamido pentanamide hydrochloride, bz-arg-nh hcl, bz-arg-nh2 hcl, benzoyl-l-arginine amide hydrochloride, n-benzoyl-l-arginine amide hydrochloride, benzoyl-arginine amide monohydrochloride monihydrate, nalpha-benzoyl-l-argininamidehydrochloridemonohydrate, 2s-5-diaminomethylidene amino-2-phenylformamido pentanamide hydrochloride
PubChem CID 22871352
IUPAC Name N-[(2S)-1-amino-5-(diaminomethylideneamino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)N.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 313.79
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Benzoyl-L-argininamide Hydrochloride Monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.