Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-DL-histidine, 98%

Click to view available options
Quantity:
5g
25g
19728-57-5
C14H15N3O4
289.291
MFCD00037852
WCOJOHPAKJFUDF-UHFFFAOYSA-N
n-cbz-dl-histidine,z-dl-his-oh,cbz-dl-histidine,2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,carbobenzoxy-dl-histidine,2-benzyloxy carbonyl amino-3-3h-imidazol-4-yl propanoic acid,n-benzyloxy carbonyl histidine,acmc-209u4b,n-carbobenzoxy-dl-histidine,.alpha.-n-cbz-dl-histidine
85793
3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O
19728-57-5
C14H15N3O4
289.291
MFCD00037852
WCOJOHPAKJFUDF-UHFFFAOYSA-N
n-cbz-dl-histidine,z-dl-his-oh,cbz-dl-histidine,2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,carbobenzoxy-dl-histidine,2-benzyloxy carbonyl amino-3-3h-imidazol-4-yl propanoic acid,n-benzyloxy carbonyl histidine,acmc-209u4b,n-carbobenzoxy-dl-histidine,.alpha.-n-cbz-dl-histidine
85793
3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O
Formula Weight 289.29
Physical Form Powder
Percent Purity 98%
Color White
Quantity 5g
Chemical Name or Material Nalpha-Benzyloxycarbonyl-DL-histidine

storageNote1 : 2° to 8°C