Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Boc-L-lysine, 97%

Catalog No. p-7055314
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 13734-28-6 | C11H22N2O4 | 246.31 g/mol

N-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13734-28-6
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00038203
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
Synonym boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine
PubChem CID 2733284
IUPAC Name (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O

Specifications

Melting Point ∼205°C (decomposition)
Color White
Beilstein 4252546
Solubility Information Soluble in water. Slightly soluble in methanol.
Quantity 5 g
Formula Weight 246.3
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-L-lysine
EINECSNumber 237-303-4
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.