Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Boc-Nepsilon-Fmoc-D-lysine, 95%

Catalog No. p-7041441
Click to view available options
Quantity:
1 g
5 g

CAS: 115186-31-7 | C26H32N2O6 | 468.55 g/mol

Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115186-31-7
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD00076966
InChI Key JYEVQYFWINBXJU-ANBDAQEENA-N
Synonym boc-d-lys fmoc-oh, n-boc-n'-fmoc-d-lysine, boc-l-lys fmoc-oh, boc-n-epsilon-fmoc-d-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-9-fluorenylmethyloxycarbonyl-d-lysine, 2r-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzbaa1040, n-boc-n-fmoc-d-lysine, pubchem12013, n2-boc-n6-fmoc-d-lysine
PubChem CID 14019424
IUPAC Name (2R)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 117°C to 123°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 468.55
Percent Purity 95%
Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-D-lysine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.