Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Fmoc-L-arginine, 95%

Catalog No. AAH6666514
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH6666514

Supplier: Thermo Scientific Chemicals H6666514

Only null left
Add to Cart

CAS: 91000-69-0 | C21H24N4O4 | 396.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91000-69-0
Molecular Formula C21H24N4O4
Molecular Weight (g/mol) 396.45
MDL Number MFCD00051770
InChI Key DVBUCBXGDWWXNY-SFHVURJKSA-N
Synonym fmoc-arg-oh, fmoc-l-arginine, nalpha-fmoc-l-arginine, alfa-fmoc-l-arginine, fmoc-l-arg-oh, n-a-fmoc-l-arginine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-guanidino-pentanoic acid, 2s-5-amidinoamino-2-fluoren-9-ylmethoxy carbonylamino pentanoic acid, 2s-5-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-arg
PubChem CID 2724631
IUPAC Name (2S)-5-{[amino(iminiumyl)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoate
SMILES NC(=[NH2+])NCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Color White
Beilstein 4828015
Quantity 25 g
Formula Weight 396.45
Sensitivity Moisture sensitive
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nalpha-Fmoc-L-arginine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.