Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine, 95%

Click to view available options
Quantity:
1g
5g
25g
120075-24-3
C31H36N4O7S
608.71
MFCD00065616
LKGHIEITYHYVED-AREMUKBSSA-N
fmoc-d-arg mtr-oh,n-fmoc-n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-d-arginine,ambotzfaa1663,fmoc-d-asp mtr-oh,fmoc-d-arg mtr-oh hplc,n-alpha-fmoc-n-omega-4-methoxy-2,3,6trimethylbenzenesulfonyl-d-arginine,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-3-4-methoxy-2,3,6-trimethylbenzenesulfonyl carbamimidamido pentanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl carbamimidamido pentanoic acid,2r-5-amino-4-methoxy-2,3,6-trimethyl-phenyl sulfonylamino methylene amino-2-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n'-4-methoxy-2,3,6-trimethylphenyl-sulfonyl-d-arginine
16211044
(2R)-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N)C)C)OC
120075-24-3
C31H36N4O7S
608.71
MFCD00065616
LKGHIEITYHYVED-AREMUKBSSA-N
fmoc-d-arg mtr-oh,n-fmoc-n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-d-arginine,ambotzfaa1663,fmoc-d-asp mtr-oh,fmoc-d-arg mtr-oh hplc,n-alpha-fmoc-n-omega-4-methoxy-2,3,6trimethylbenzenesulfonyl-d-arginine,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-3-4-methoxy-2,3,6-trimethylbenzenesulfonyl carbamimidamido pentanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl carbamimidamido pentanoic acid,2r-5-amino-4-methoxy-2,3,6-trimethyl-phenyl sulfonylamino methylene amino-2-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n'-4-methoxy-2,3,6-trimethylphenyl-sulfonyl-d-arginine
16211044
(2R)-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N)C)C)OC
Formula Weight 608.71
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine

storageNote1 : 2° to 8°C