Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha,Nepsilon-Di-Fmoc-D-lysine, 98%

Click to view available options
Quantity:
250mg
1g
5g
75932-02-4
C36H34N2O6
590.676
MFCD00270743
BMJRTKDVFXYEFS-MGBGTMOVSA-N
fmoc-d-lys fmoc-oh,r-2,6-bis 9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,2r-2,6-bis 9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzfaa1331,n2,n6-bis-fmoc-d-lysine,n-a,n-e-di-fmoc-d-lysine,n-a,n-epsilon-di-fmoc-d-lysine,n,n'-bis-9-fluorenylmethyloxycarbonyl-d-lysine,n-alpha-n-epsilon-bis 9-fluorenylmethyloxycarbonyl-d-lysine,n~2~,n~6~-bis 9h-fluoren-9-yl methoxy carbonyl-d-lysine
22816294
(2R)-2,6-bis(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
75932-02-4
C36H34N2O6
590.676
MFCD00270743
BMJRTKDVFXYEFS-MGBGTMOVSA-N
fmoc-d-lys fmoc-oh,r-2,6-bis 9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,2r-2,6-bis 9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzfaa1331,n2,n6-bis-fmoc-d-lysine,n-a,n-e-di-fmoc-d-lysine,n-a,n-epsilon-di-fmoc-d-lysine,n,n'-bis-9-fluorenylmethyloxycarbonyl-d-lysine,n-alpha-n-epsilon-bis 9-fluorenylmethyloxycarbonyl-d-lysine,n~2~,n~6~-bis 9h-fluoren-9-yl methoxy carbonyl-d-lysine
22816294
(2R)-2,6-bis(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
Formula Weight 590.68
Physical Form Solid
Percent Purity 98%
Quantity 250mg
Chemical Name or Material Nalpha,Nepsilon-Di-Fmoc-D-lysine

storageNote1 : 2° to 8°C