Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-Nepsilon-carbobenzoxy-L-lysine 98.0+%, TCI America™
SDP

Catalog No. B163225G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2389-45-9
Molecular Formula C19H28N2O6
Molecular Weight (g/mol) 380.44
MDL Number MFCD00065584
InChI Key BDHUTRNYBGWPBL-GGYSOQFKNA-N
Synonym boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine
PubChem CID 2724765
IUPAC Name (2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 77°C
Color White
Quantity 25 g
Formula Weight 380.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-Nepsilon-carbobenzoxy-L-lysine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.