Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-tele-(p-toluenesulfonyl)-L-histidine 98.0+%, TCI America™
SDP

Catalog No. B206625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B206625G 25 g
B20665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B206625G Supplier TCI America Supplier No. B206625G

Chemical Identifiers

CAS 35899-43-5
Molecular Formula C18H23N3O6S
Molecular Weight (g/mol) 409.46
MDL Number MFCD00065967
InChI Key DCLJSEPKYJSEHW-UHFFFAOYNA-N
Synonym boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh
PubChem CID 6455010
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3-[1-(4-methylbenzenesulfonyl)-1H-imidazol-4-yl]propanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Formula Weight 409.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-tele-(p-toluenesulfonyl)-L-histidine
EINECSNumber (9)-0933
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.