Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144487 Supplier Invitrogen™ Supplier No. PA5144487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NAM8 Polyclonal Antibody for Western Blot, ELISA

Component of the U1 small nuclear ribonucleoprotein complex (U1 snRNP) involved in the initiation of meiotic recombination (PubMed:1603056, PubMed:7625279). Involved in the formation of DSBs at recombination hot-spots through meiosis-specific splicing of REC107 pre-mRNA (PubMed:7625279, PubMed:9112441, PubMed:21788335). Collaborates with MER1 to promote splicing of essential meiotic mRNAs REC10, AMA1, MER3, HFM1, SPO22 and PCH2 (PubMed:10983980, PubMed:21208980, PubMed:21788335). NAM8 interacts with the pre-mRNA downstream of the 5' splice site, in a region of non-conserved sequence and is required for efficient splicing of uncapped RNA precursor (PubMed:10072385).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAM8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene NAM8
Gene Accession No. Q00539
Gene Alias MRE2; MUD15
Gene Symbols NAM8
Host Species Rabbit
Immunogen Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) NAM8 protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856486
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.