Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAMPT Monoclonal Antibody (C8), Invitrogen™

Catalog No. FEMA550952
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550952 20 μL
FEMA550951 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550952 Supplier Invitrogen™ Supplier No. MA550952
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAMPT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene NAMPT
Gene Accession No. P43490, Q52I78, Q99KQ4
Gene Alias 1110035O14Rik; AI314458; AI480535; DKFZp666B131; EC 2.4.2.12; hypothetical protein; MGC117256; NAmPRTase; NAMPT; nampt1; nampta; NAMPTP1; Nicotinamide phosphoribosyltransferase; nicotinamide phosphoribosyltransferase 1; nicotinamide phosphoribosyltransferase a; nicotinamide phosphoribosyltransferase pseudogene 1; PBEF; Pbef1; pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; pre-B-cell colony-enhancing factor 1; Pre-B-cell colony-enhancing factor 1 homolog; pre-B-cell colony-enhancing factor 1 precursor; VF; VISFATIN
Gene Symbols NAMPT
Host Species Mouse
Immunogen Recombinant Human VF protein, Met1-His491 (Accession #P43490).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10135, 59027, 595123
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.