Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAMPT, Mouse anti-Human, Clone: OMNI379, Abnova™

Catalog No. 89998818 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-818 Supplier Abnova Corporation Supplier No. MAB16026
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen NAMPT
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OMNI379
Conjugate Unconjugated
Dilution ELISA (1:2000-10000) Flow Cytometry (1:1000) Immunocytochemistry (1:500-1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-500) Immunoprecipitation (1:500-1000) Western Blot (1:2000-5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4.
Gene nicotinamide phosphoribosyltransferase
Gene Alias 1110035O14Rik/DKFZp666B131/MGC117256/PBEF/PBEF1/VF/VISFATIN
Gene Symbols NAMPT
Host Species Mouse
Immunogen Recombinant protein corresponding to human NAMPT.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10135
Target Species Human, Mouse, Rat
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.