Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAMPT/PBEF Mouse anti-Human, Mouse, Rat, Clone: 3D4D8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NAMPT/PBEF |
---|---|
Clone | 3D4D8 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-114-2755
![]() ![]() |
Proteintech Group Inc
663851IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.Specifications
NAMPT/PBEF | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
1110035O14Rik, NAmPRTase, NAMPT, PBEF, PBEF1, Pre B cell enhancing factor, VF, VISFATIN | |
Nampt | |
IgG2b | |
Protein A | |
Antibody |
3D4D8 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P43490, Q80Z29, Q99KQ4 | |
10135, 297508, 59027 | |
NAMPT/PBEF Fusion Protein Ag2434 | |
Primary | |
-20°C | |
NAMPT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title