Learn More
NAMPT/PBEF/Visfatin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300372AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat and Pig
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.Specifications
NAMPT/PBEF/Visfatin | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P43490, Q99KQ4 | |
NAMPT | |
Between 400 and 450 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
NAMPT | |
NAMPT, Nicotinamide phosphoribosyltransferase, EC 2.4.2.12, NAmPRTase, Nampt, Pre-B cell-enhancing factor, Pre-B-cell colony-enhancing factor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10135, 59027 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.