Learn More
NAMPT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA585532
Description
Keep as concentrated solution. Positive Control: NCI-H929, Neuro 2A, C8D30, NIH-3T3, Raw264.7, mouse muscle, Rat2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also involved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.
Specifications
NAMPT | |
Polyclonal | |
Unconjugated | |
NAMPT | |
VF Antibody; nicotinamide phosphoribosyltransferase Antibody; PBEF Antibody; 1110035O14Rik Antibody; VISFATIN Antibody; PBEF1 Antibody | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
59027, 10135, 297508 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q99KQ4, Q80Z29, P43490 | |
Nampt | |
Recombit protein encompassing a sequence within the N-terminus region of human Visfatin. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.