Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Nano-Glo™ Luciferase Assay System

Catalog No. PRN1130 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
10 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1130 10 x 10 mL
PRN1150 10 x 100 mL
PRN1110 10 mL
PRN1120 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRN1130 Supplier Promega Supplier No. N1130
Add to Cart
Add to Cart

The Nano-Glo™ Luciferase Assay System provides a simple, single-addition reagent that generates a glow-type signal in the presence of NanoLuc luciferase with a half-life of approximately 120 minutes in commonly used tissue culture media.

  • Add-Mix-Measure assay for NanoLuc(R) Luciferase
  • Bright NanoLuc(R) reporter enables use in applications where sensitivity is limited
  • Add-and-read simplicity allows scaling from bench to HTS
  • Extended bright light output optimized for batch and continuous processing

Specifications

Kit Contents 10 x 200μL Nano-Glo™ Luciferase assay Substrate, 10 x 10mL Nano-Glo™ Luciferase Assay Buffer, integral Lysis Buffer reagent
Type Nano-Glo Luciferase Assay Buffer
Quantity 10 x 10 mL
Content And Storage −30 to −10°C
For Use With (Application) Provides a single addition reagent that generates a glow-type signal in the presence of NanoLuc™ luciferase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.