Learn More
Promega NanoBiT PPI Flexi Starter System, 1 each
Catalog No. PRN2015 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRN2015 1 Each
1 options
Catalog No. PRN2015 Supplier Promega Supplier No. N2015
The NanoBiT PPI Starter Systems provide the vectors required to create the LgBiT and SmBiT protein fusions, a PRKACA:PRKAR2A positive control pair and a negative control vector. Starter systems also include the Nano-Glo Live Cell Assay System.
  • Sensitive Detection of Protein Interactions in Live Cells
  • Detects interactions at low expression levels and allows real−time kinetic analysis in live cells
  • Reversible protein complementation reporter to analyze protein association and disassociation
  • Improved sensitivity and accuracy over split firefly luciferase
Specifications
Content And Storage -30°C to -10°C
Quantity 1 Each