Learn More
Promega NanoBRET™ Nano-Glo™ Substrate
Catalog No. PRN1571 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 μL
5 x 50 μL
2 x 1.25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1571 50 μL
PRN1572 5 x 50 μL
PRN1573 2 x 1.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRN1571 Supplier Promega Supplier No. N1571
Add to Cart
Add to Cart
The NanoBRET™ Nano-Glo(R) Substrate is the substrate used by NanoLuc(R) Luciferase to generate the donor signal in several NanoBRET™ Assays.
For use with NanoBRET assay, a proximity-based assay that can detect protein interactions by measuring energy transfer from a bioluminescent protein donor to a fluorescent protein acceptor.
Specifications
Content And Storage -30°C to -10°C
Quantity 50 μL