Learn More
Promega NanoBRET™ PPI MCS Starter System
Catalog No. prn1811
Click to view available options
Quantity:
1/Ea.
Protein:Protein Interaction (PPI) Assays use NanoLuc Luciferase and HaloTag™ protein fused to target proteins of interest to enable sensitive, reproducible detection of protein interactions in the natural cellular environment.
  • BRET-Based Assays to Detect Protein:Protein Interactions in Live Cells
  • Study induction and inhibition of protein interactions using full-length proteins expressed at physiologically relevant levels
  • Starter Systems provide fusion vectors, positive controls and detection reagents for building PPI assays
  • Choice of multicloning site (MCS) or Flexi(R) vector-based cloning options
Specifications
Content And Storage COMPONENTS AT DIFFERENT TEMPS
Quantity 1/Ea.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.