Learn More
Promega NanoBRET TE Intracellular BET BRD Assay, 1,000 assays
Catalog No. PRN2131 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRN2131 1000 Assays
1 options
Catalog No. PRN2131 Supplier Promega Supplier No. N2131
Add to Cart
Add to Cart
Request a Quote
NanoBRET Target Engagement Intracellular BET BRD Assay measures compound binding at select BET bromodomain target proteins within intact cells. The assay is based on NanoBRET, an energy transfer technique for measuring molecular proximity in living cells.
  • Directly Measure BET BRD−Test Compound Affinity and Residence Time
  • Compound and tracer compete directly for the binding site
  • Assays rely on full−length proteins that are similar to the native forms
  • Express target proteins at low levels
Specifications
Content And Storage LESS THAN -65°C
Quantity 1000 Assays