Learn More
Promega NanoBRET TE Intracellular BET BRD Assay, 100 assays
Catalog No. PRN2130 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 assays
Catalog No. Quantity
PRN2130 100 assays
1 options
Catalog No. PRN2130 Supplier Promega Supplier No. N2130
NanoBRET Target Engagement Intracellular BET BRD Assay measures compound binding at select BET bromodomain target proteins within intact cells. The assay is based on NanoBRET, an energy transfer technique for measuring molecular proximity in living cells.
  • Directly Measure BET BRD−Test Compound Affinity and Residence Time
  • Compound and tracer compete directly for the binding site
  • Assays rely on full−length proteins that are similar to the native forms
  • Express target proteins at low levels
Specifications
Content And Storage LESS THAN -65C
Quantity 100 assays