Learn More
Promega NanoBRET TE Intracellular BET BRD Complete Kit, 1,000 assays
Catalog No. PRN2180 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRN2180 1000 Assays
1 options
Catalog No. PRN2180 Supplier Promega Supplier No. N2180
Add to Cart
Add to Cart
Request a Quote
NanoBRET Target Engagement Intracellular BET BRD Assay measures compound binding at select BET bromodomain target proteins within intact cells. The assay is based on NanoBRET, an energy transfer technique for measuring molecular proximity in living cells.
  • Directly Measure BET BRD−Test Compound Affinity and Residence Time
  • Compound and tracer compete directly for the binding site
  • Assays rely on full−length proteins that are similar to the native forms
  • Express target proteins at low levels
Specifications
Quantity 1000 Assays