Learn More
Promega NanoBRET TE Intracellular BET BRD Detection Reagents, 10,000 assays
Catalog No. PRN2140 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
Catalog No. Quantity
PRN2140 10,000 Assays
1 options
Catalog No. PRN2140 Supplier Promega Supplier No. N2140
Add to Cart
Add to Cart
Request a Quote
NanoBRET Target Engagement Intracellular BET BRD Assay measures compound binding at select BET bromodomain target proteins within intact cells. The assay is based on NanoBRET, an energy transfer technique for measuring molecular proximity in living cells.
  • Directly Measure BET BRD−Test Compound Affinity and Residence Time
  • Compound and tracer compete directly for the binding site
  • Assays rely on full−length proteins that are similar to the native forms
  • Express target proteins at low levels
Specifications
Content And Storage LESS THAN -65°C
Quantity 10,000 Assays