Learn More
Promega NanoBRET TE Intracellular HDAC Assay, 100 assays
Catalog No. PRN2080 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 Assays
Catalog No. Quantity
PRN2080 100 Assays
1 options
Catalog No. PRN2080 Supplier Promega Supplier No. N2080
NanoBRET Target Engagement Intracellular HDAC Assay measures compound binding at select HDAC target proteins within intact cells. The assay is based on NanoBRET System, an energy transfer technique designed to measure molecular proximity in living cells.
  • Directly Measure HDAC−Test Compound Affinity and Residence Time in Live Cells
  • Compound and tracer compete directly for the binding site
  • Assays rely on full−length proteins that are similar to the native forms
  • Express target proteins at low levels
Specifications
Content And Storage LESS THAN -65°C
Quantity 100 Assays