Learn More
Promega NanoBRET TE Intracellular HDAC Complete Kit, 1,000 assays
Catalog No. PRN2170 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRN2170 1000 Assays
1 options
Catalog No. PRN2170 Supplier Promega Supplier No. N2170
Add to Cart
Add to Cart
Request a Quote
NanoBRET Target Engagement Intracellular HDAC Assay measures compound binding at select HDAC target proteins within intact cells. The assay is based on NanoBRET System, an energy transfer technique designed to measure molecular proximity in living cells.
  • Directly Measure HDAC−Test Compound Affinity and Residence Time in Live Cells
  • Compound and tracer compete directly for the binding site
  • Assays rely on full−length proteins that are similar to the native forms
  • Express target proteins at low levels
Specifications
Quantity 1000 Assays