Learn More
Promega NanoBRET TE Intracellular Kinase Detection Reagents, K-5, 10,000 assays
Catalog No. PRN2530
Click to view available options
Quantity:
10,000 Assays
The NanoBRET Target Engagement (TE) Assay measures compound binding at select kinase targets within intact cells. The assay is based on the NanoBRET System, an energy transfer technique designed to measure molecular proximity in living cells.
  • Measure Compound Binding at Kinase Targets in Intact Cells
  • Detect kinase target engagement in live cells
  • Choose from >125 kinase fusion vectors
  • See Kinase Table (below) for available kinases and related information
Specifications
Quantity 10,000 Assays
Storage Requirements <−65°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.