Learn More
Therapak™ NanoCool™ Refrigerated Specimen Shipper
No gel refrigerants required
$1032.00 - $1478.00
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
22-130-418
|
Therapak™
56850 |
Sturdy transport box | Up to 36 standard 13 x 100mm tubes |
Case of 2 for $1,032.00
|
|
||||
22-130-417
|
Therapak™
56849 |
Transport box | Up to 18 standard 13 x 100mm tubes |
Case of 4 for $1,478.00
|
|
||||
Description
- Portable unit maintains temperature from 2-8°C — ideal for overnight or international shipments
- Weighs less and requires less space than conventional polystyrene boxes and ice packs, reducing shipping costs
- Vacuum Insulated Panel (VIP) compartment transfers heat by evaporating small quantities of water at low pressure; a heat absorption method that's seven times more efficient than ice
- Separate NanoCool cooler fits atop the VIP compartment and can be activated at room temperature with the push of a button
- Small shipper holds up to 18 standard (13 × 100mm) transport tubes; large shipper holds up to 36 tubes
Order Info
One-time use only; cannot be reconditioned
Specifications
Portable Unit | |
For overnight or international shipping |