Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ Lite Spectrophotometer, NanoDrop Lite
SCI_ED

Catalog No. s07018
Click to view available options
Detector Type:
Silicon Photodiode

The Thermo Scientific™ NanoDrop Lite Spectrophotometer is a compact, personal UV-Vis microvolume spectrophotometer that complements the full-featured NanoDrop 2000/2000c and NanoDrop 8000 instruments.

The NanoDrop Lite performs basic microvolume measurements. Its compact design, with built-in controls and software, make the NanoDrop Lite small enought to fit on any benchtop. The patented sample retention system allows sample to be pipetted directly onto the optical measurement surface. After measurement the sample is wiped off the measurement surface with a lint free lab wipe.
  • Measures nucleic acid concentration at 260nm and purity using the 260/280 ratio
  • Measures purified protein concentration at 280nm
  • Employs the unique NanoDrop microvolume sampling technology
  • Delivers the accuracy and reproducibility expected from NanoDrop instruments
  • Uses built-in controls and software–no computer required
  • Optional accessory printer for cryogenic labels available
  • TRUSTED_SUSTAINABILITY

    Specifications

    Absorbance Accuracy 0.03
    Absorbance Range 0 to 30 A
    Certifications/Compliance UL/CSA and CE
    Compatibility Local Control Only
    Description NanoDrop Lite UV-Vis Spectrophotometer
    Detection Limits 0.12 mg/mL to 45 mg/mL (BSA), 4 ng/μ
    Detector Type Silicon Photodiode
    Display LCD
    Light Source LED
    Voltage 6 V
    Footprint 16 x 11.5 cm
    For Use With (Application) Nucleic acid, protein A280
    Measurement Time <5 sec.
    No. of Samples 1
    Pathlength 0.5 mm
    Sample Volume (Metric) 1 to 2 μL
    Spectral Resolution <8.0 nm
    Volume (Metric) 1 μL
    Warranty 1 year parts and labor
    Wattage 18 W
    Wavelength Repeatability 0.002 *SD of 10 individual measurements at 0.74 Abs
    Weight (Metric) 0.8 kg
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.