Learn More
Thermo Scientific™ NanoDrop™ One/OneC IQ/OQ Kits
Description
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ). Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Solutions include:
- Information on the validation process
- Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
- Information on pharmacopoeia recommended tests for specific instrument performance where applicable
- Installation and instrument certification
Solutions available for GENESYS 40-180, BioMate 160, Evolution200 series, Evolution 350, NanoDrop One, NanoDrop 2000, and NanoDrop 8000 instruments.
IQ/OQ Document Kit includes:
- USB stick with IQ/OQ procedure
- Qualification Binder Kit (A4 or US)
- CHEM-PR1-KIT,
- CHEM-PV-1
Ensures your instrument is installed correctly and working within manufacturer's specifications.
Specifications
Specifications
| Description | NanoDrop One IQ/OQ Kit -US |
| Type | IQ/OQ Kit (US) |
| For Use With (Equipment) | NanoDrop One/OneC Spectrophotometers |
| Includes | IQ/OQ procedure (USB), Qualification Binder Kit (US), CHEM-PR1-KIT, and CHEM-PV-1 |