Learn More
Thermo Scientific™ NanoDrop One Microvolume UV-Vis Spectrophotometer PROMO
Description
Our next generation Thermo Scientific™ NanoDrop™ One microvolume spectrophotometers are pushing the boundaries once again with the Thermo Scientific™ Acclaro™ Sample Intelligence Software that helps you undersatnd the quality of your sample beore you use it in downstream applications, bringing you one step closer to success.
Product Features:
- Full spectrum microvolume UV-Vis spectrophotometer with 190 nm to 850 nm wavelength measurement range
- Sample vol 1.0 to 2.0 μL
- Measurement range 2 to 27500 ng/μL dsDNA
- Column integrity sensor
- Tilting and sliding screen
- 2 yr warranty
Quantify and qualify DNA, RNA, and protein samples with only 1 to 2 μL and have full-spectral data in seconds using the Thermo Scientific™ NanoDrop™ One Microvolume UV-Vis spectrophotometer.
- Prevent costly delays - identify contaminants, obtain corrected concentrations, and enjoy success in your downstream applications
- Collect data fast - Auto-Blank and Auto-Measure modes save time while organizing data
- Save bench space - user-friendly local control with sleek, standalone design means no PC required
Thermo Scientific™ Acclaro™ Sample Intelligence Software enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting.
- Verify sample quality in seconds - identify sample contaminants and obtain corrected concentration results
- Access information through alerts - receive instant feedback about sample quality with on-demand technical support and guided troubleshooting
- Rely on accurate results - ensure measurement integrity with embedded sensor and digital image analysis
NanoDrop One Enhancements
- Ergonomic, standalone design - integrated Android™ tablet eliminates the need for a separate computer and provides modern connectivity for seamless data transfer via Wireless, Ethernet or USB.
- Auto-Measure and Auto-Blank functions - multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
- Wider dynamic range - no dilutions required for highly concentrated samples (dsDNA at 27,500 ng/μL) with auto-range pathlength technology.
- Integrated Learning Center - an archive of answers with technical support documents and educational animations at your fingertips.NanoDrop One/Onec Spectrophotometer capabilities:
- Wide spectral range (190 to 850 nm) for measuring a variety of samples types:
- Peptides (205 nm)
- DNA and RNA (260 nm)
- Purified protein (280 nm)
- Toxicology assays and industrial dyes (490 nm)
- Gold nanoparticles (520 nm)
- Colorimetric protein assays (BCA 562 nm, Bradford 595 nm, Modified Lowry 650 nm, Pierce 660 660 nm)
- Optical Density measurements (600 nm)
- Combining the patented sample-retention system with cuvette capability accommodates low and high concentrations (2.0 - 27,500 ng/μL dsDNA and 0.06 - 820 mg/mL BSA)
- Pedestal measurements that require only 0.5 - 2 μL of your sample and do not require sample dilution, even for highly concentrated samples
- Pre-configured methods for DNA, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, and Lowry
- User-friendly software that includes custom methods and data export capabilities
- Instrument comes with a 2-year warranty for worry-free support

Specifications
Specifications
Includes | PR-1 pedestal conditioning solution, PV-1 performance verification solution, Microfiber screen wipe, USA Power Cable |
Warranty | 2 Years |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.