Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ One Microvolume UV-Vis Spectrophotometer, Trade-up Promotion

Catalog No. 13400518LY
Click to view available options
Warranty:
2 Years

Currently own a Thermo Scientific™ NanoDrop™? Take advantage of this loyalty trade-up promotion. Proof of existing unit with serial number may be requested.

Our next generation Thermo Scientific™ NanoDrop™ One microvolume spectrophotometers are pushing the boundaries once again with the Thermo Scientific™ Acclaro™ Sample Intelligence Software that helps you understand the quality of your sample beore you use it in downstream applications, bringing you one step closer to success.

Product Features:

  • Full spectrum microvolume UV-Vis spectrophotometer with 190nm-850nm wavelength measurement range
  • Sample vol 1.0 - 2.0 μL
  • Measurement range 2 - 27500 ng/μL dsDNA
  • Column integrity sensor
  • Tilting and sliding screen
  • 2 yr warranty

Quantify and qualify DNA, RNA, and protein samples with only 1-2 μL and have full-spectral data in seconds using the Thermo Scientific™ NanoDrop™ One Microvolume UV-Vis spectrophotometer.

  • Prevent costly delays — identify contaminants, obtain corrected concentrations, and enjoy success in your downstream applications
  • Collect data fast — Auto-Blank and Auto-Measure modes save time while organizing data
  • Save bench space — user-friendly local control with sleek, standalone design means no PC required

Thermo Scientific™ Acclaro™ Sample Intelligence Software enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting.

  • Verify sample quality in seconds — identify sample contaminants and obtain corrected concentration results
  • Access information through alerts — receive instant feedback about sample quality with on-demand technical support and guided troubleshooting
  • Rely on accurate results — ensure measurement integrity with embedded sensor and digital image analysis

NanoDrop One Enhancements

  • Ergonomic, standalone design — integrated Android™ tablet eliminates the need for a separate computer and provides modern connectivity for seamless data transfer via Wireless*, Ethernet or USB.
  • Auto-Measure and Auto-Blank functions — multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
  • Wider dynamic range — no dilutions required for highly concentrated samples (dsDNA at 27,500 ng/μL) with auto-range pathlength technology.
  • Integrated Learning Center — an archive of answers with technical support documents and educational animations at your fingertips.

NanoDrop One/Onec Spectrophotometer capabilities:

  • Wide spectral range (190-850 nm) for measuring a variety of samples types:
    • Peptides (205 nm)
    • DNA and RNA (260 nm)
    • Purified protein (280 nm)
    • Toxicology assays and industrial dyes (490 nm)
    • Gold nanoparticles (520 nm)
    • Colorimetric protein assays (BCA 562 nm, Bradford 595 nm, Modified Lowry 650 nm, Pierce 660 660 nm)
    • Optical Density measurements (600 nm)
  • Combining the patented** sample-retention system with cuvette capability accommodates low and high concentrations (2.0 - 27,500 ng/μL dsDNA and 0.06 - 820 mg/mL BSA)
  • Pedestal measurements that require only 0.5 - 2 μL of your sample and do not require sample dilution, even for highly concentrated samples
  • Pre-configured methods for DNA, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, and Lowry
  • User-friendly software that includes custom methods and data export capabilities
  • Instrument comes with a 2-year warranty for worry-free support

* Wi-Fi model not available in certain countries - consult your local NanoDrop distributor

**Patents US6628382 and US6809826

Android is a trademark of Google Inc.

For research use only. Not for diagnostic purposes.

All NanoDrop instruments are approved to CE and UL/CSA.

a
TRUSTED_SUSTAINABILITY

Specifications

Includes PR-1 pedestal conditioning solution, PV-1 performance verification solution, Microfiber screen wipe, USA Power Cable
Warranty 2 Years
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.