Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ NanoDrop™ One/OneC Productivity Kits
Click to view available options
Type:
NanoDrop One Productivity Kit
NanoDrop OneC Productivity Kit
Description
The Thermo Scientific™ NanoDrop™ One and NanoDrop™ OneC Productivity Kits allow you to improve the productivity of your NanoDrop One and NanoDrop OneC Microvolume UV-Vis Spectrophotometers.
The NanoDrop One Productivity Kit includes:
- NanoDrop-branded Finnpipette™ F1 Adjustable-Volume Pipette , 0.2–2 μL
- Microfiber screen wipe for NanoDrop One Spectrophotometer
- NanoDrop PR-1 Reconditioning Compound Kit
- NanoDrop PV-1 Performance Verification Solution
- I love my NanoDrop sticker
The NanoDrop OneC Productivity Kit includes:
- NanoDrop-branded Finnpipette™ F1 Adjustable Volume Pipette , 0.2–2μL
- Microfiber screen wipe for NanoDrop OneC spectrophotometer
- NanoDrop PR-1 Reconditioning Compound Kit
- NanoDrop PV-1 Performance Verification Solution
- 10mm path quartz cuvette
- Cuvette stir bars, 5x2mm, pack of 10
- I love my NanoDrop sticker
For Research Use Only. Not for use in diagnostic procedures.

Specifications
Specifications
Type | NanoDrop One Productivity Kit |
For Use With (Equipment) | NanoDrop One |
Includes |
|
Storage Requirements | Store at room temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction