Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ NanoDrop™ OneC Microvolume UV-Vis Spectrophotometer PROMO
Click to view available options
Warranty:
2 Years
Description
Our next generation Thermo Scientific™ NanoDrop™ OneC microvolume spectrophotometers are pushing the boundaries once again with the Thermo Scientific™ Acclaro™ Sample Intelligence Software that helps you understand the quality of your sample before you use it in downstream applications, bringing you one step closer to success. NanoDrop™ OneC spectrophotometer features a thermostated, stirred cuvette position for additional life science measurements.
Product Features:
Quantify and qualify DNA, RNA, and protein samples with only 1-2μL and have full-spectral data in seconds using the Thermo Scientific™ NanoDrop™ OneC Microvolume UV-Vis spectrophotometer.
NanoDrop OneC Instrument Highlights
Thermo Scientific™ Acclaro™ Sample Intelligence Software enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting.
NanoDrop One/OneC Spectrophotometer capabilities:
* Wi-Fi model not available in certain countries - consult your local NanoDrop distributor
**Patents US6628382 and US6809826
Android is a trademark of Google Inc.
For research use only. Not for diagnostic purposes.
All NanoDrop instruments are approved to CE and UL/CSA.
Product Features:
- Full spectrum microvolume UV-Vis spectrophotometer with cuvette position (stirring, 37°C thermostatting, kinetics)
- Meas. range 0.4 - 27500ng/μL dsDNA
- 190nm-850nm wavelength meas range
- Column integrity sensor
- 2 yr warranty
Quantify and qualify DNA, RNA, and protein samples with only 1-2μL and have full-spectral data in seconds using the Thermo Scientific™ NanoDrop™ OneC Microvolume UV-Vis spectrophotometer.
- Prevent costly delays — identify contaminants, obtain corrected concentrations, and enjoy success in your downstream applications
- Collect data fast — Auto-Blank and Auto-Measure modes save time while organizing data
- Save bench space — user-friendly local control with sleek, standalone design means no PC required
NanoDrop OneC Instrument Highlights
- Contains both pedestal and cuvette positions to expand experimental flexibility and increase the dynamic range.
- Measure dilute samples, perform kinetics experiments and take optical density measurements of bacterial cultures.
- Cuvette position includes temperature control and stirring.
Thermo Scientific™ Acclaro™ Sample Intelligence Software enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting.
- Verify sample quality in seconds — identify sample contaminants and obtain corrected concentration results
- Access information through alerts — receive instant feedback about sample quality with on-demand technical support and guided troubleshooting
- Rely on accurate results — ensure measurement integrity with embedded sensor and digital image analysis
- Ergonomic, standalone design — integrated Android™ tablet eliminates the need for a separate computer and provides modern connectivity for seamless data transfer via Wireless*, Ethernet or USB.
- Auto-Measure and Auto-Blank functions — multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
- Wider dynamic range — no dilutions required for highly concentrated samples (dsDNA at 27,500ug/μL) with auto-range pathlength technology.
- Integrated Learning Center — an archive of answers with technical support documents and educational animations at your fingertips.
NanoDrop One/OneC Spectrophotometer capabilities:
- Wide spectral range (190-850 nm) for measuring a variety of samples types:
- Peptides (205 nm)
- DNA and RNA (260 nm)
- Purified protein (280 nm)
- Toxicology assays and industrial dyes (490 nm)
- Gold nanoparticles (520 nm)
- Colorimetric protein assays (BCA 562 nm, Bradford 595 nm, Modified Lowry 650 nm, Pierce 660 660 nm)
- Optical Density measurements (600 nm)
- Combining the patented** sample-retention system with cuvette capability accommodates low and high concentrations (2.0 - 27,500ng/ μL dsDNA and 0.06 - 820mg/mL BSA)
- Pedestal measurements that require only 0.5 - 2μL of your sample and do not require sample dilution, even for highly concentrated samples
- Pre-configured methods for DNA, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, and Lowry
- User-friendly software that includes custom methods and data export capabilities
- Instrument comes with a 2-year warranty for worry-free support
* Wi-Fi model not available in certain countries - consult your local NanoDrop distributor
**Patents US6628382 and US6809826
Android is a trademark of Google Inc.
For research use only. Not for diagnostic purposes.
All NanoDrop instruments are approved to CE and UL/CSA.

Specifications
Specifications
Includes | PR-1 pedestal conditioning solution, PV-1 performance verification solution, USB memory device, Microfiber screen wipe, USA Power Cable |
Warranty | 2 Years |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction