Learn More
Thermo Scientific NanoDrop™ Reagents
Catalog No. p-10341000
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
Certified standards for verifying instrument performance to factory specifications are available separately.
For use with Thermo Scientific NanoDrop Spectrophotometers.
  • Ten times more concentrated compared to other K2Cr2O7 calibration solutions
  • Packaged in 0.5mL snap-top glass ampules
  • Intended for single use only
TRUSTED_SUSTAINABILITY
Specifications
Type CF-1 Calibration Check Fluid
For Use With (Equipment) NanoDrop 8000, NanoDrop Lite, NanoDrop 2000, NanoDrop 2000c spectrophotometers
Packaging 0.5 mL snap-top glass ampules
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.